missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Keto-4,5,6,7-tetrahydrothianaphthene, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC122890100
Specifications
4-Keto-4,5,6,7-tetrahydrothianaphthene | |
Authentic | |
1.5895 to 1.5915 | |
White to Yellow | |
10g | |
96.0 | |
96% min. (GC) | |
MFCD00005861 | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
6,7-dihydro-5H-1-benzothiophen-4-one | |
83418 | |
Low Melting Solid or Liquid After Melting |
110°C | |
Glass bottle | |
120°C (10.0mmHg) | |
34°C to 40°C | |
13414-95-4 | |
100.0 | |
C8H8OS | |
4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro | |
C1CC2=C(C=CS2)C(=O)C1 | |
152.21 | |
152.21 | |
97% |
Chemical Identifiers
13414-95-4 | |
152.21 | |
GJEKNELSXNSYAQ-UHFFFAOYSA-N | |
83418 | |
C1CC2=C(C=CS2)C(=O)C1 |
C8H8OS | |
MFCD00005861 | |
4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro | |
6,7-dihydro-5H-1-benzothiophen-4-one |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 236-510-7