missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Keto-4,5,6,7-tetrahydrothianaphthene, 97%, ACROS Organics™

$98.40 - $98.40

Chemical Identifiers

CAS 13414-95-4
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.21
MDL Number MFCD00005861
InChI Key GJEKNELSXNSYAQ-UHFFFAOYSA-N
Synonym 4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro
PubChem CID 83418
IUPAC Name 6,7-dihydro-5H-1-benzothiophen-4-one
SMILES C1CC2=C(C=CS2)C(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122890010
SDS View Documents
Thermo Fisher Scientific
122890010
1g Glass bottle
Each for $98.40
 
AC122890100
SDS View Documents
Thermo Fisher Scientific
AC122890100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13414-95-4
152.21
GJEKNELSXNSYAQ-UHFFFAOYSA-N
83418
C1CC2=C(C=CS2)C(=O)C1
C8H8OS
MFCD00005861
4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro
6,7-dihydro-5H-1-benzothiophen-4-one

Specifications

110°C
Glass bottle
120°C (10.0mmHg)
34°C to 40°C
13414-95-4
100.0
C8H8OS
4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro
C1CC2=C(C=CS2)C(=O)C1
152.21
152.21
97%
Authentic
1.5895 to 1.5915
White to Yellow
1g
96.0
96% min. (GC)
MFCD00005861
GJEKNELSXNSYAQ-UHFFFAOYSA-N
6,7-dihydro-5H-1-benzothiophen-4-one
83418
Low Melting Solid or Liquid After Melting
4-Keto-4,5,6,7-tetrahydrothianaphthene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-510-7

SDS