missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Keto-4,5,6,7-tetrahydrothianaphthene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 122890010

 View more versions of this product

Catalog No. AC122890010



Description

Description

Specifications

Specifications

4-Keto-4,5,6,7-tetrahydrothianaphthene
Authentic
1.5895 to 1.5915
White to Yellow
1g
96.0
96% min. (GC)
MFCD00005861
GJEKNELSXNSYAQ-UHFFFAOYSA-N
6,7-dihydro-5H-1-benzothiophen-4-one
83418
Low Melting Solid or Liquid After Melting
110°C
Glass bottle
120°C (10.0mmHg)
34°C to 40°C
13414-95-4
100.0
C8H8OS
4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro
C1CC2=C(C=CS2)C(=O)C1
152.21
152.21
97%

Chemical Identifiers

13414-95-4
152.21
GJEKNELSXNSYAQ-UHFFFAOYSA-N
83418
C1CC2=C(C=CS2)C(=O)C1
C8H8OS
MFCD00005861
4,5,6,7-tetrahydro-4-benzothiophenone, 4,5,6,7-tetrahydrothionaphthen-4-one, 4-keto-4,5,6,7-tetrahydrothianaphthene, 4-oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-dihydro-1-benzothiophen-4 5h-one, 6,7-dihydro-4-benzo b thiophenone, 6,7-dihydro-5h-benzo b thiophen-4-one, 6,7-dihydrobenzo b thiophen-4 5h-one, 6,7-dihydrobenzo b thiophen-4-one, benzo b thiophen-4 5h-one, 6,7-dihydro
6,7-dihydro-5H-1-benzothiophen-4-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-510-7

SDS Documents