missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical 97.0+%, TCI America™

$246.84 - $1,463.34

Chemical Identifiers

CAS 36410-81-8
Molecular Formula C10H17N2OS
Molecular Weight (g/mol) 213.319
MDL Number MFCD00010109
InChI Key BKENZAFIBRCMKD-UHFFFAOYSA-N
Synonym 1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl, 4-isothiocyanato-1, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical, 4-isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxy radical, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxyl, 4-isothiocyanato-tempo, 4-isothiocyanato-tempo, for esr-spectroscopy, 4-itc-tempo
PubChem CID 147645
IUPAC Name 4-isothiocyanato-1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine
SMILES CC1(CC(CC(N1[O])(C)C)N=C=S)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0486100MG View Documents Tci America
I0486100MG
100mg Each for $246.84
 
I04861G View Documents Tci America
I04861G
1g Each for $1,463.34
 
Specifications

Chemical Identifiers

36410-81-8
213.319
BKENZAFIBRCMKD-UHFFFAOYSA-N
147645
CC1(CC(CC(N1[O])(C)C)N=C=S)C
C10H17N2OS
MFCD00010109
1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl, 4-isothiocyanato-1, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical, 4-isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxy radical, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxyl, 4-isothiocyanato-tempo, 4-isothiocyanato-tempo, for esr-spectroscopy, 4-itc-tempo
4-isothiocyanato-1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine

Specifications

Undesignated
100mg
C10H17N2OS
1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl, 4-isothiocyanato-1, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical, 4-isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxy radical, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxyl, 4-isothiocyanato-tempo, 4-isothiocyanato-tempo, for esr-spectroscopy, 4-itc-tempo
CC1(CC(CC(N1[O])(C)C)N=C=S)C
213.319
213.32
≥97.0% (GC,T)
128°C
36410-81-8
MFCD00010109
BKENZAFIBRCMKD-UHFFFAOYSA-N
4-isothiocyanato-1-$l^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine
147645
Crystal-Powder at 20°C
4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-Oxyl Free Radical
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer