missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropylthiazole-2-carboxylic acid, 97%

$280.66 - $280.66

Chemical Identifiers

CAS 300831-06-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD09864275
InChI Key RDWHKWXYJQUZNS-UHFFFAOYSA-N
Synonym 4-isopropylthiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, 4-1-methylethyl, 4-isopropyl-1,3-thiazole-2-carboxylic acid, 4-1-methylethyl-2-thiazolecarboxylicacid, 4-isopropyl-thiazole-2-carboxylic acid, 4-propan-2-yl-1,3-thiazole-2-carboxylic acid, 4-1-methylethyl-2-thiazolecarboxylic acid, pubchem19253, 4-isopropylthiazole-2-carboxlic acid, 4-isopropyl-thiazole-2-carboxlic acid
PubChem CID 22499307
IUPAC Name 4-propan-2-yl-1,3-thiazole-2-carboxylic acid
SMILES CC(C)C1=CSC(=N1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6621603 View Documents Alfa Aesar™
H6621603
1g Each for $280.66
 
Specifications

Chemical Identifiers

300831-06-5
171.214
RDWHKWXYJQUZNS-UHFFFAOYSA-N
22499307
CC(C)C1=CSC(=N1)C(=O)O
C7H9NO2S
MFCD09864275
4-isopropylthiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, 4-1-methylethyl, 4-isopropyl-1,3-thiazole-2-carboxylic acid, 4-1-methylethyl-2-thiazolecarboxylicacid, 4-isopropyl-thiazole-2-carboxylic acid, 4-propan-2-yl-1,3-thiazole-2-carboxylic acid, 4-1-methylethyl-2-thiazolecarboxylic acid, pubchem19253, 4-isopropylthiazole-2-carboxlic acid, 4-isopropyl-thiazole-2-carboxlic acid
4-propan-2-yl-1,3-thiazole-2-carboxylic acid

Specifications

1g
C7H9NO2S
14226164
RDWHKWXYJQUZNS-UHFFFAOYSA-N
4-propan-2-yl-1,3-thiazole-2-carboxylic acid
22499307
Solid
4-Isopropylthiazole-2-carboxylic acid
300831-06-5
MFCD09864275
4-isopropylthiazole-2-carboxylic acid, 2-thiazolecarboxylic acid, 4-1-methylethyl, 4-isopropyl-1,3-thiazole-2-carboxylic acid, 4-1-methylethyl-2-thiazolecarboxylicacid, 4-isopropyl-thiazole-2-carboxylic acid, 4-propan-2-yl-1,3-thiazole-2-carboxylic acid, 4-1-methylethyl-2-thiazolecarboxylic acid, pubchem19253, 4-isopropylthiazole-2-carboxlic acid, 4-isopropyl-thiazole-2-carboxlic acid
CC(C)C1=CSC(=N1)C(=O)O
171.214
171.22
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning