missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylpyridine, TCI America™

$554.59 - $1,630.17

Chemical Identifiers

CAS 696-30-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00039719
InChI Key FRGXNJWEDDQLFH-UHFFFAOYSA-N
PubChem CID 69674
IUPAC Name 4-propan-2-ylpyridine
SMILES CC(C)C1=CC=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I01745G View Documents TCI America
I01745G
5g Each for $554.59
 
I017425G View Documents TCI America
I017425G
25g Each for $1,630.17
 
Specifications

Chemical Identifiers

696-30-0
121.183
FRGXNJWEDDQLFH-UHFFFAOYSA-N
4-propan-2-ylpyridine
C8H11N
MFCD00039719
69674
CC(C)C1=CC=NC=C1

Specifications

178°C
5g
C8H11N
FRGXNJWEDDQLFH-UHFFFAOYSA-N
4-propan-2-ylpyridine
69674
Clear Liquid at 20°C
4-Isopropylpyridine
Yellow
696-30-0
MFCD00039719
CC(C)C1=CC=NC=C1
121.183
121.18
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : Yes