missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylpiperidine, TCI America™

$291.81 - $1,011.41

Chemical Identifiers

CAS 19678-58-1
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD09055342
InChI Key YBPWIUSXQXYTSR-UHFFFAOYSA-N
PubChem CID 140569
IUPAC Name 4-propan-2-ylpiperidine
SMILES CC(C)C1CCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I07335G View Documents TCI America
I07335G
5g Each for $1,011.41
 
I07331G View Documents TCI America
I07331G
1g Each for $291.81
 
Specifications

Chemical Identifiers

19678-58-1
127.231
YBPWIUSXQXYTSR-UHFFFAOYSA-N
4-propan-2-ylpiperidine
C8H17N
MFCD09055342
140569
CC(C)C1CCNCC1

Specifications

171°C
5g
C8H17N
1993
CC(C)C1CCNCC1
127.231
127.23
≥98.0% (GC,T)
Yellow
19678-58-1
MFCD09055342
YBPWIUSXQXYTSR-UHFFFAOYSA-N
4-propan-2-ylpiperidine
140569
Clear Liquid at 20°C
4-Isopropylpiperidine
Safety and Handling

Safety and Handling

TSCA : No