missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropylphenyl isothiocyanate, 96%

Manufacturer:  Alfa Aesar™ L1294904

 View more versions of this product

Catalog No. AAL1294904



Specifications

Specifications

4-Isopropylphenyl isothiocyanate
2g
C10H11NS
UN2922
4-isopropylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanatobenzene, 4-isopropylphenylisothiocyanate, 1-isothiocyanato-4-propan-2-yl benzene, 4-methylethyl benzenisothiocyanate, acmc-20amft, 4isopropylphenylisothiocyanate, 4-iso-propylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanato-benzene, 1-isopropyl-4-isothiocyanatobenzene #
CC(C)C1=CC=C(C=C1)N=C=S
177.265
177.27
154°C to 155°C (20mmHg)
89007-45-4
MFCD00022060
3240731
SNWJJTIJIDXIHL-UHFFFAOYSA-N
1-isothiocyanato-4-propan-2-ylbenzene
145194
96%

Chemical Identifiers

89007-45-4
177.265
SNWJJTIJIDXIHL-UHFFFAOYSA-N
145194
CC(C)C1=CC=C(C=C1)N=C=S
C10H11NS
MFCD00022060
4-isopropylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanatobenzene, 4-isopropylphenylisothiocyanate, 1-isothiocyanato-4-propan-2-yl benzene, 4-methylethyl benzenisothiocyanate, acmc-20amft, 4isopropylphenylisothiocyanate, 4-iso-propylphenyl isothiocyanate, 1-isopropyl-4-isothiocyanato-benzene, 1-isopropyl-4-isothiocyanatobenzene #
1-isothiocyanato-4-propan-2-ylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents