Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropylphenyl isocyanate, 98%

Chemical Identifiers

Chemical Name or Material 4-Isopropylphenyl isocyanate
CAS 31027-31-3
Molecular Formula C10H11NO
Formula Weight 161.21
MDL Number MFCD00013880
UN Number UN2206
Beilstein 2936403
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1121704

  SDS
alfa aesar™
L1121704
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1121709

  SDS
alfa aesar™
L1121709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.21
Chemical Name or Material 4-Isopropylphenyl isocyanate
Assay 98%
Molecular Formula C10H11NO
MDL Number MFCD00013880
CAS 31027-31-3
UN Number UN2206
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Boiling Point 84° to 85°C (4mmHg)
Beilstein 2936403
Flash Point 90°C (194°F)
Quantity 2g
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C