missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylphenyl isocyanate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1121709

 View more versions of this product

Catalog No. AAL1121709



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Isopropylphenyl isocyanate
1.01
90°C (194°F)
1.5205
10 g
2936403
4-isopropylphenyl isocyanate, 4-isopropylphenylisocyanate, 1-isocyanato-4-isopropylbenzene, 1-isocyanato-4-isopropyl-benzene, p-isopropylphenyl isocyanate, benzene, 1-isocyanato-4-1-methylethyl, 4-isopropyl phenylisocyanate, 4-methylethyl benzenisocyanate, p-cumenyl isocyanate, acmc-1aild
CC(C)C1=CC=C(C=C1)N=C=O
161.204
161.21
31027-31-3
84°C to 85°C (4 mmHg)
C10H11NO
MFCD00013880
UN2206
Moisture sensitive
PJVRNNRZWASOIT-UHFFFAOYSA-N
1-isocyanato-4-propan-2-ylbenzene
94416
98%

Chemical Identifiers

31027-31-3
161.204
PJVRNNRZWASOIT-UHFFFAOYSA-N
94416
CC(C)C1=CC=C(C=C1)N=C=O
C10H11NO
MFCD00013880
4-isopropylphenyl isocyanate, 4-isopropylphenylisocyanate, 1-isocyanato-4-isopropylbenzene, 1-isocyanato-4-isopropyl-benzene, p-isopropylphenyl isocyanate, benzene, 1-isocyanato-4-1-methylethyl, 4-isopropyl phenylisocyanate, 4-methylethyl benzenisocyanate, p-cumenyl isocyanate, acmc-1aild
1-isocyanato-4-propan-2-ylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P284-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330-H334

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 250-439-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only