Learn More
4-Isopropylphenol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 174510010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Isopropylphenol | |
99-89-8 | |
100.0 | |
Authentic | |
Glass bottle | |
(CH3)2CHC6H4OH | |
1kg | |
4-isopropylphenol, p-isopropylphenol, p-cumenol, australol, phenol, 4-1-methylethyl, 4-1-methylethyl phenol, 1-hydroxy-4-isopropylbenzene, 4-propan-2-yl phenol, phenol, p-isopropyl, para-isopropylphenol | |
YQUQWHNMBPIWGK-UHFFFAOYSA-N | |
4-propan-2-ylphenol | |
7465 | |
98% |
98% | |
97.5 | |
212.0°C to 212.4°C | |
97.5% min. (GC) | |
C9H12O | |
MFCD00002372 | |
06,505 | |
Solubility in water: .. Other solubilities: soluble in 10% sodium hydroxide,soluble in alcohol and ethers | |
CC(C)C1=CC=C(C=C1)O | |
136.19 | |
136.19 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 202-798-8
RTECSNumber : SL5950000
TSCA : TSCA
RUO â Research Use Only