missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropylbiphenyl, 97%

$46.80 - $143.78

Chemical Identifiers

CAS 7116-95-2
Molecular Formula C15H16
Formula Weight 196.29
MDL Number MFCD00221718
Beilstein 1858288
Chemical Name or Material 4-Isopropylbiphenyl
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1440503 SDS Alfa Aesar™
L1440503
1g Each for $46.80
AAL1440506 SDS Alfa Aesar™
L1440506
5g Each for $143.78
Specifications Description & Specifications

Chemical Identifiers

7116-95-2
196.29
1858288
C15H16
MFCD00221718
4-Isopropylbiphenyl

Specifications

110° to 112°C (1mmHg)
1g
C15H16
MFCD00221718
4-Isopropylbiphenyl
11°C
7116-95-2
196.29
1858288
97%

Safety and Handling

RTECSNumber : DV5415000