missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropylbenzyl chloride, 95%

Manufacturer:  Alfa Aesar™ L1183906

 View more versions of this product

Catalog No. AAL1183906



Specifications

Specifications

4-Isopropylbenzyl chloride
Pungent
5g
C10H13Cl
UN3265
4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl
CC(C)C1=CC=C(C=C1)CCl
168.664
168.67
104°C (219°F)
119°C to 120°C (35mmHg)
2051-18-5
MFCD00018885
907207
CYAKWEQUWJAHLW-UHFFFAOYSA-N
1-(chloromethyl)-4-propan-2-ylbenzene
74916
95%

Chemical Identifiers

2051-18-5
168.664
CYAKWEQUWJAHLW-UHFFFAOYSA-N
74916
CC(C)C1=CC=C(C=C1)CCl
C10H13Cl
MFCD00018885
4-isopropylbenzyl chloride, 1-chloromethyl-4-isopropylbenzene, p-cymene, 7-chloro, p-isopropylbenzyl chloride, 7-chloro-p-cymene, 4-isopropylbenzylchloride, benzene, 1-chloromethyl-4-1-methylethyl, 1-chloromethyl-4-propan-2-yl benzene, p-isopropyl benzyl chloride, benzene, 1-chloromethyl-4-methylethyl
1-(chloromethyl)-4-propan-2-ylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents