missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzyl bromide, 97%, ACROS Organics™

Chemical Identifiers

CAS 73789-86-3
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
InChI Key YXTHBZLABLYGEE-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-isopropylbenzene, 4-isopropyl benzyl bromide, 4-isopropylbenzyl bromide, 4-isopropylbenzylbromide, benzene, 1-bromomethyl-4-1-methylethyl, bromo-p-cymeme, p-cymene, bromo, p-isopropylbenzyl bromide
PubChem CID 47719
IUPAC Name 1-(bromomethyl)-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC438480010
View Documents
Thermo Fisher Scientific
AC438480010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

73789-86-3
213.118
1-bromomethyl-4-isopropylbenzene, 4-isopropyl benzyl bromide, 4-isopropylbenzyl bromide, 4-isopropylbenzylbromide, benzene, 1-bromomethyl-4-1-methylethyl, bromo-p-cymeme, p-cymene, bromo, p-isopropylbenzyl bromide
1-(bromomethyl)-4-propan-2-ylbenzene
C10H13Br
YXTHBZLABLYGEE-UHFFFAOYSA-N
47719
CC(C)C1=CC=C(C=C1)CBr

Specifications

Authentic
Colorless to Yellow
73789-86-3
100.0
C10H13Br
YXTHBZLABLYGEE-UHFFFAOYSA-N
1-(bromomethyl)-4-propan-2-ylbenzene
47719
Liquid
4-Isopropylbenzyl bromide
Glass bottle
1g
96.0
97%
1-bromomethyl-4-isopropylbenzene, 4-isopropyl benzyl bromide, 4-isopropylbenzyl bromide, 4-isopropylbenzylbromide, benzene, 1-bromomethyl-4-1-methylethyl, bromo-p-cymeme, p-cymene, bromo, p-isopropylbenzyl bromide
CC(C)C1=CC=C(C=C1)CBr
213.118
213.12
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger