missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isopropylbenzyl alcohol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC187805000

 View more versions of this product

Catalog No. AC187805000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Isopropylbenzyl alcohol
>112°C
Glass bottle
0.98
Colorless to Yellow
500g
96.0
96% min. (GC)
(CH3)2CHC6H4CH2OH
06, III, 1911
4-isopropylbenzyl alcohol, 4-isopropylphenyl methanol, cumic alcohol, cumin alcohol, cuminic alcohol, cuminol, cuminyl alcohol, cumyl alcohol, p-cymen-7-ol, p-isopropylbenzyl alcohol
CC(C)C1=CC=C(C=C1)CO
150.22
CHEBI:27628
Liquid
0.9800g/mL
Authentic
1.5180 to 1.52
248°C
28°C
536-60-7
100.0
C10H14O
MFCD00004663
15, 2609
OIGWAXDAPKFNCQ-UHFFFAOYSA-N
(4-propan-2-ylphenyl)methanol
325
150.22
97%

Chemical Identifiers

536-60-7
150.22
OIGWAXDAPKFNCQ-UHFFFAOYSA-N
325
(4-propan-2-ylphenyl)methanol
C10H14O
MFCD00004663
4-isopropylbenzyl alcohol, 4-isopropylphenyl methanol, cumic alcohol, cumin alcohol, cuminic alcohol, cuminol, cuminyl alcohol, cumyl alcohol, p-cymen-7-ol, p-isopropylbenzyl alcohol
CHEBI:27628
CC(C)C1=CC=C(C=C1)CO
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 208-640-4

SDS Documents