Please login to your online account to display your discounted pricing

4-Isopropylbenzyl alcohol, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  187800250

 View more versions of this product

Catalog No. AC187800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 150.22
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 06, III, 1911
Merck Index 15, 2609
Density 0.98g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Isopropylbenzyl alcohol
Infrared Spectrum Authentic
Molecular Formula C10H14O
Linear Formula (CH3)2CHC6H4CH2OH
CAS 536-60-7
Packaging Glass bottle
Refractive Index 1.5180 to 1.5200 (20°C, 589nm)
Assay Percent Range 97%
MDL Number MFCD00004663
Synonym Cumic alcohol
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Isopropylbenzyl alcohol
CAS 536-60-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H14O
Linear Formula (CH3)2CHC6H4CH2OH
Formula Weight 150.22
MDL Number MFCD00004663
Synonym Cumic alcohol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents