Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropylbenzoic acid, 99%

$64.49 - $798.58

Chemical Identifiers

Chemical Name or Material 4-Isopropylbenzoic acid
CAS 536-66-3
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00002564
Merck Index 14,2618
Synonym Cuminic acid; Cumic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2070406

  SDS
alfa aesar™
B2070406
5g Each for $64.49

AAB2070414

  SDS
alfa aesar™
B2070414
25g Each for $247.11

AAB2070422

  SDS
alfa aesar™
B2070422
100g Each for $798.58
Description & Specifications

Specifications

Formula Weight 164.2
Quantity 5g
CAS 536-66-3
MDL Number MFCD00002564
Assay 99%
Synonym Cuminic acid; Cumic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Merck Index 14,2618
Molecular Formula C10H12O2
RTECS Number DH3850500
TSCA TSCA
Melting Point 117° to 120°C
Chemical Name or Material 4-Isopropylbenzoic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.