Please login to your online account to display your discounted pricing

4-isopropylbenzenesulfonyl chloride, 94%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Isopropylbenzenesulfonyl chloride
CAS 54997-90-9
CAS Min % 92.0
CAS Max % 100.0
Assay 92% min.
Assay Percent Range 94%
Molecular Formula C9H11ClO2S
Linear Formula (CH3)2CHC6H4SO2Cl
Formula Weight 218.70g/mol
MDL Number MFCD00041510
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376100050

  SDS
thermo fisher scientific
376100050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.2250g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5380 to 1.5410 (20°C, 589nm)
Color Clear yellow to orange
Quantity 5g
Chemical Name or Material 4-Isopropylbenzenesulfonyl chloride
CAS 54997-90-9
CAS Min % 92.0
CAS Max % 100.0
Physical Form Clear yellow to orange liquid
Assay 92% min.
Assay Percent Range 94%
Molecular Formula C9H11ClO2S
Linear Formula (CH3)2CHC6H4SO2Cl
Formula Weight 218.70g/mol
MDL Number MFCD00041510
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.