missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Isopropylbenzeneboronic acid, 98+%, Thermo Scientific Chemicals
$182.42 - $720.93
Chemical Identifiers
CAS | 16152-51-5 |
---|---|
Molecular Formula | C9H13BO2 |
Molecular Weight (g/mol) | 164.011 |
MDL Number | MFCD01074614 |
InChI Key | IAEUFBDMVKQCLU-UHFFFAOYSA-N |
Synonym | 4-isopropylphenylboronic acid, 4-isopropylbenzeneboronic acid, 4-isopropylphenyl boronic acid, 4-propan-2-ylphenyl boronic acid, p-isopropylphenylboronic acid, p-cumylboronic acid, 4-cumylboronic acid, 4-iso-propylphenyl boronic acid, 4-iso-propylphenylboronic acid |
PubChem CID | 2773480 |
IUPAC Name | (4-propan-2-ylphenyl)boronic acid |
SMILES | B(C1=CC=C(C=C1)C(C)C)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1745903
|
Thermo Scientific Chemicals
L1745903 |
1 g |
N/A
|
|
|||||
AAL1745906
|
Thermo Scientific Chemicals
L1745906 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16152-51-5 | |
164.011 | |
IAEUFBDMVKQCLU-UHFFFAOYSA-N | |
2773480 | |
B(C1=CC=C(C=C1)C(C)C)(O)O |
C9H13BO2 | |
MFCD01074614 | |
4-isopropylphenylboronic acid, 4-isopropylbenzeneboronic acid, 4-isopropylphenyl boronic acid, 4-propan-2-ylphenyl boronic acid, p-isopropylphenylboronic acid, p-cumylboronic acid, 4-cumylboronic acid, 4-iso-propylphenyl boronic acid, 4-iso-propylphenylboronic acid | |
(4-propan-2-ylphenyl)boronic acid |
Specifications
16152-51-5 | |
C9H13BO2 | |
1 g | |
4-isopropylphenylboronic acid, 4-isopropylbenzeneboronic acid, 4-isopropylphenyl boronic acid, 4-propan-2-ylphenyl boronic acid, p-isopropylphenylboronic acid, p-cumylboronic acid, 4-cumylboronic acid, 4-iso-propylphenyl boronic acid, 4-iso-propylphenylboronic acid | |
B(C1=CC=C(C=C1)C(C)C)(O)O | |
164.011 | |
164.01 | |
4-Isopropylbenzeneboronic acid |
142°C to 144°C | |
MFCD01074614 | |
2830262 | |
IAEUFBDMVKQCLU-UHFFFAOYSA-N | |
(4-propan-2-ylphenyl)boronic acid | |
2773480 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only