missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Isopropylacetophenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2058106
Description
4'-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
4'-Isopropylacetophenone | |
0.97 | |
C11H14O | |
MFCD00048297 | |
2205694 | |
Soluble in alcohol. Insoluble in water. | |
CC(C)C1=CC=C(C=C1)C(C)=O | |
162.23 | |
162.23 |
645-13-6 | |
238°C | |
1.523 | |
5 g | |
4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one | |
PDLCCNYKIIUWHA-UHFFFAOYSA-N | |
1-(4-propan-2-ylphenyl)ethanone | |
12578 | |
98% |
Chemical Identifiers
645-13-6 | |
162.23 | |
PDLCCNYKIIUWHA-UHFFFAOYSA-N | |
12578 | |
CC(C)C1=CC=C(C=C1)C(C)=O |
C11H14O | |
MFCD00048297 | |
4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one | |
1-(4-propan-2-ylphenyl)ethanone |
Safety and Handling
EINECSNumber : 211-433-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only