Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Isopropyl)thiophenol, 95%

Chemical Identifiers

Chemical Name or Material (4-Isopropyl)thiophenol
CAS 4946-14-9
Molecular Formula C9H12S
Formula Weight 152.26
MDL Number MFCD00041505
UN Number UN1760
Beilstein 1906198
Synonym 4-Isopropylbenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1603806

  SDS
alfa aesar™
A1603806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1603814

  SDS
alfa aesar™
A1603814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.26
Chemical Name or Material (4-Isopropyl)thiophenol
Assay 95%
Molecular Formula C9H12S
MDL Number MFCD00041505
CAS 4946-14-9
UN Number UN1760
Beilstein 1906198
Synonym 4-Isopropylbenzenethiol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Flash Point 106°C (222°F)
Odor Stench
Boiling Point 99° to 100°C (14mmHg)
Quantity 5g

Applications
(4-Isopropyl)thiophenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.