Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropyl-beta-nitrostyrene, 97%

$110.61 - $427.92

Chemical Identifiers

Chemical Name or Material 4-Isopropyl-beta-nitrostyrene
CAS 42139-37-7
Molecular Formula C11H13NO2
Formula Weight 191.23
MDL Number MFCD00085100
Synonym 1-(4-Isopropylphenyl)-2-nitroethylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1095214

  SDS
alfa aesar™
A1095214
25g Each for $427.92

AAA1095206

  SDS
alfa aesar™
A1095206
5g Each for $110.61
Description & Specifications

Specifications

Formula Weight 191.23
Quantity 25g
Chemical Name or Material 4-Isopropyl-beta-nitrostyrene
Melting Point 35° to 38°C
Molecular Formula C11H13NO2
MDL Number MFCD00085100
CAS 42139-37-7
Synonym 1-(4-Isopropylphenyl)-2-nitroethylene
Assay 97%