Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropyl-3-methylaniline hydrochloride, 97%

$102.31 - $402.31

Chemical Identifiers

Chemical Name or Material 4-Isopropyl-3-methylaniline hydrochloride
CAS 4534-11-6
Molecular Formula C10H15N·HCl
Formula Weight 185.7
MDL Number MFCD00156562
Synonym 4-Isopropyl-m-toluidine hydrochloride; 3-Methyl-4-isopropylaniline hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1975406

  SDS
alfa aesar™
A1975406
5g Each for $102.31

AAA1975414

  SDS
alfa aesar™
A1975414
25g Each for $402.31
Description & Specifications

Specifications

Formula Weight 185.7
Chemical Name or Material 4-Isopropyl-3-methylaniline hydrochloride
Assay 97%
Molecular Formula C10H15N·HCl
MDL Number MFCD00156562
CAS 4534-11-6
Synonym 4-Isopropyl-m-toluidine hydrochloride; 3-Methyl-4-isopropylaniline hydrochloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 222° to 225°C
Quantity 5g

Applications
4-Isopropyl-3-methylaniline hydrochloride is used to synthesis of fine chemical, organic intermediates.

Solubility
Slightly soluble in water (9.9 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.