Please login to your online account to display your discounted pricing

4-Isopropoxyphenylboronic acid, 97%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393930100

 View more versions of this product

Catalog No. AC393930100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 180.01
Quantity 10g
Color White to off-white or light tan
CAS Max % 100.0
CAS Min % 96.0
Linear Formula (CH3)2CHOC6H4B(OH)2
Packaging Glass bottle
Molecular Formula C9H13BO3
CAS 153624-46-5
Chemical Name or Material 4-Isopropoxyphenylboronic acid
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03427051
Synonym p-Isopropoxybenzeneboronic acid
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 4-Isopropoxyphenylboronic acid
CAS 153624-46-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H13BO3
Linear Formula (CH3)2CHOC6H4B(OH)2
Formula Weight 180.01
MDL Number MFCD03427051
Synonym p-Isopropoxybenzeneboronic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents