Please login to your online account to display your discounted pricing

4-Isopropoxyphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Isopropoxyphenylboronic acid
CAS 153624-46-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H13BO3
Linear Formula (CH3)2CHOC6H4B(OH)2
Formula Weight 180.01
MDL Number MFCD03427051
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC393930010

  SDS
thermo fisher scientific
AC393930010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC393930100

  SDS
thermo fisher scientific
AC393930100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 180.01
Quantity 1g
Color White to off-white or light tan
CAS Min % 96.0
CAS Max % 100.0
Synonym p-Isopropoxybenzeneboronic acid
Chemical Name or Material 4-Isopropoxyphenylboronic acid
Assay Percent Range 97%
Molecular Formula C9H13BO3
CAS 153624-46-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Linear Formula (CH3)2CHOC6H4B(OH)2
MDL Number MFCD03427051
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.