Please login to your online account to display your discounted pricing

4-Isopropoxycarbonylphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Isopropoxycarbonylphenylboronic acid
Name Note 97%
CAS 342002-82-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H13BO4
Formula Weight 208.02
MDL Number MFCD02259385
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369410050

  SDS
thermo fisher scientific
AC369410050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC369410010

  SDS
thermo fisher scientific
AC369410010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 208.02
Quantity 5g
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Isopropoxycarbonylphenylboronic acid
Assay Percent Range 97%
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 342002-82-8
Infrared Spectrum Authentic
Name Note 97%
Molecular Formula C10H13BO4
MDL Number MFCD02259385
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage Refrigerator +4°C
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Refrigerator +4°C