missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Isopropoxycarbonyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5333506

 View more versions of this product

Catalog No. AAH5333506



Description

Description

Applications
4-(Isopropoxycarbonyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-(Isopropoxycarbonyl)benzeneboronic acid
5g
C10H13BO4
4-isopropoxycarbonylphenylboronic acid, 4-isopropoxycarbonyl phenyl boronic acid, 4-isopropoxycarbonyl benzeneboronic acid, propan-2-yl 4-dihydroxyboranyl benzoate, isopropyl 4-boronobenzoate, 4-isopropoxycarbonyl phenylboronic acid, p-isopropoxycarbonyl phenylboronic acid, 4-isopropoxycarbonylphenylboronicacid, 4-propan-2-yloxycarbonylphenyl boronic acid, acmc-209i5o
B(C1=CC=C(C=C1)C(=O)OC(C)C)(O)O
208.02
208.02
111°C
342002-82-8
MFCD02259385
NIQHFKWRHJZMQU-UHFFFAOYSA-N
(4-propan-2-yloxycarbonylphenyl)boronic acid
2734703
98%

Chemical Identifiers

342002-82-8
208.02
NIQHFKWRHJZMQU-UHFFFAOYSA-N
2734703
B(C1=CC=C(C=C1)C(=O)OC(C)C)(O)O
C10H13BO4
MFCD02259385
4-isopropoxycarbonylphenylboronic acid, 4-isopropoxycarbonyl phenyl boronic acid, 4-isopropoxycarbonyl benzeneboronic acid, propan-2-yl 4-dihydroxyboranyl benzoate, isopropyl 4-boronobenzoate, 4-isopropoxycarbonyl phenylboronic acid, p-isopropoxycarbonyl phenylboronic acid, 4-isopropoxycarbonylphenylboronicacid, 4-propan-2-yloxycarbonylphenyl boronic acid, acmc-209i5o
(4-propan-2-yloxycarbonylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents