missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isopropoxy-2-methylbenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5313203

 View more versions of this product

Catalog No. AAH5313203



Specifications

Specifications

4-Isopropoxy-2-methylbenzeneboronic acid
1g
C10H15BO3
4-isopropoxy-2-methylphenylboronic acid, 4-isopropoxy-2-methylphenyl boronic acid, 4-isopropoxy-2-methylbenzeneboronic acid, boronic acid,b-2-methyl-4-1-methylethoxy phenyl, 2-methyl-4-propan-2-yloxy phenyl boronic acid, acmc-209qf7, 4-isopropoxy-2-methylphenyl boronicacid, 4-isopropoxy-2-methyl-phenyl boronic acid, 2-methyl-4-propan-2-yl oxy phenyl boronic acid
B(C1=C(C=C(C=C1)OC(C)C)C)(O)O
194.037
194.04
112°C to 116°C
871126-21-5
MFCD07363788
OEBXBUNQDNBWRP-UHFFFAOYSA-N
(2-methyl-4-propan-2-yloxyphenyl)boronic acid
16218044
98%

Chemical Identifiers

871126-21-5
194.037
OEBXBUNQDNBWRP-UHFFFAOYSA-N
16218044
B(C1=C(C=C(C=C1)OC(C)C)C)(O)O
C10H15BO3
MFCD07363788
4-isopropoxy-2-methylphenylboronic acid, 4-isopropoxy-2-methylphenyl boronic acid, 4-isopropoxy-2-methylbenzeneboronic acid, boronic acid,b-2-methyl-4-1-methylethoxy phenyl, 2-methyl-4-propan-2-yloxy phenyl boronic acid, acmc-209qf7, 4-isopropoxy-2-methylphenyl boronicacid, 4-isopropoxy-2-methyl-phenyl boronic acid, 2-methyl-4-propan-2-yl oxy phenyl boronic acid
(2-methyl-4-propan-2-yloxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents