missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isocyanatobenzoyl chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 3729-21-3
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.575
InChI Key JZXYEMZTQVSTJC-UHFFFAOYSA-N
Synonym acmc-20amg0, 4-isocyanato benzoyl chloride, 4-isocyanatobenzoic acid chloride, 4-isocyanatobenzoyl chloride, benzoyl chloride, 4-isocyanato-9ci
PubChem CID 2733323
IUPAC Name 4-isocyanatobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC314420050 View Documents Acros Organics
314420050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC314420010 View Documents Acros Organics
AC314420010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

3729-21-3
181.575
acmc-20amg0, 4-isocyanato benzoyl chloride, 4-isocyanatobenzoic acid chloride, 4-isocyanatobenzoyl chloride, benzoyl chloride, 4-isocyanato-9ci
4-isocyanatobenzoyl chloride
C8H4ClNO2
JZXYEMZTQVSTJC-UHFFFAOYSA-N
2733323
C1=CC(=CC=C1C(=O)Cl)N=C=O

Specifications

Authentic
Yellow
5g
97.5
97%
acmc-20amg0, 4-isocyanato benzoyl chloride, 4-isocyanatobenzoic acid chloride, 4-isocyanatobenzoyl chloride, benzoyl chloride, 4-isocyanato-9ci
C1=CC(=CC=C1C(=O)Cl)N=C=O
181.575
181.58
≥97.5%
Glass Bottle
35°C to 38°C
3729-21-3
100.0
C8H4ClNO2
JZXYEMZTQVSTJC-UHFFFAOYSA-N
4-isocyanatobenzoyl chloride
2733323
Crystalline Mass
4-Isocyanatobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger