Please login to your online account to display your discounted pricing

4-Isocyanatobenzoyl chloride, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC314420010

 View more versions of this product

Catalog No. AC314420010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 181.58
Physical Form Crystalline Mass
Assay ≥97.5%
Quantity 1g
Color Light yellow
Melting Point 35° to 38°C
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 97%
Molecular Formula C8H4ClNO2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
Chemical Name or Material 4-Isocyanatobenzoyl chloride
CAS 3729-21-3
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.

Specifications

Chemical Name or Material 4-Isocyanatobenzoyl chloride
CAS 3729-21-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 97%
Molecular Formula C8H4ClNO2
Formula Weight 181.58
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.

SDS & Safety Documents