missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Isocyanato-3-methyl-5-phenylisoxazole, 90%, Maybridge

Chemical Identifiers

CAS 352018-89-4
Molecular Formula C11H8N2O2
Molecular Weight (g/mol) 200.197
MDL Number MFCD03086123
InChI Key ZSAPULDWGBONCI-UHFFFAOYSA-N
Synonym 4-isocyanato-3-methyl-5-phenylisoxazole, 3-methyl-5-phenylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3-methyl-5-phenyl, 3-methyl-5-phenylisoxazol-4-isocyanate, 3-methyl-5-phenylisoxazol-4-ylisocyanate, isoxazole, 4-isocyanato-3-methyl-5-phenyl-9ci
PubChem CID 2776517
IUPAC Name 4-isocyanato-3-methyl-5-phenyl-1,2-oxazole
SMILES CC1=NOC(=C1N=C=O)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC20306DA SDS View Documents Fisher Scientific Company
CC20306DA
1g Amber Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

352018-89-4
200.197
ZSAPULDWGBONCI-UHFFFAOYSA-N
2776517
CC1=NOC(=C1N=C=O)C2=CC=CC=C2
C11H8N2O2
MFCD03086123
4-isocyanato-3-methyl-5-phenylisoxazole, 3-methyl-5-phenylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3-methyl-5-phenyl, 3-methyl-5-phenylisoxazol-4-isocyanate, 3-methyl-5-phenylisoxazol-4-ylisocyanate, isoxazole, 4-isocyanato-3-methyl-5-phenyl-9ci
4-isocyanato-3-methyl-5-phenyl-1,2-oxazole

Specifications

Amber Glass Bottle
352018-89-4
100.0
C11H8N2O2
4-isocyanato-3-methyl-5-phenylisoxazole, 3-methyl-5-phenylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3-methyl-5-phenyl, 3-methyl-5-phenylisoxazol-4-isocyanate, 3-methyl-5-phenylisoxazol-4-ylisocyanate, isoxazole, 4-isocyanato-3-methyl-5-phenyl-9ci
CC1=NOC(=C1N=C=O)C2=CC=CC=C2
200.197
200.2
4-Isocyanato-3-methyl-5-phenylisoxazole
1g
89.5
90%
MFCD03086123
ZSAPULDWGBONCI-UHFFFAOYSA-N
4-isocyanato-3-methyl-5-phenyl-1,2-oxazole
2776517
90%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning