missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Isobutyramidobenzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53147MD

 View more versions of this product

Catalog No. AAH53147MD



Description

Description

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Solubility
Slightly Soluble (3.3 g/L) (25°C)

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

4-Isobutyramidobenzeneboronic acid
250mg
C10H14BNO3
4-isobutyramidobenzeneboronic acid, 4-isobutyramidophenyl boronic acid, 4-isobutyramidophenylboronic acid, 4-isobutyramido benzeneboronic acid, boronic acid,b-4-2-methyl-1-oxopropyl amino phenyl, 4-2-methylpropanamido phenylboronic acid, acmc-209qle, 4-isobutyramidophenyl boronicacid, 4-2-methylpropanamido phenyl boronic acid, 4-2-methylpropanoylamino phenyl boronic acid
B(C1=CC=C(C=C1)NC(=O)C(C)C)(O)O
207.036
207.04
235°C
874219-50-8
MFCD08235068
DBWRRIVRDITVFV-UHFFFAOYSA-N
[4-(2-methylpropanoylamino)phenyl]boronic acid
44119813
98%

Chemical Identifiers

874219-50-8
207.036
DBWRRIVRDITVFV-UHFFFAOYSA-N
44119813
B(C1=CC=C(C=C1)NC(=O)C(C)C)(O)O
C10H14BNO3
MFCD08235068
4-isobutyramidobenzeneboronic acid, 4-isobutyramidophenyl boronic acid, 4-isobutyramidophenylboronic acid, 4-isobutyramido benzeneboronic acid, boronic acid,b-4-2-methyl-1-oxopropyl amino phenyl, 4-2-methylpropanamido phenylboronic acid, acmc-209qle, 4-isobutyramidophenyl boronicacid, 4-2-methylpropanamido phenyl boronic acid, 4-2-methylpropanoylamino phenyl boronic acid
[4-(2-methylpropanoylamino)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents