missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Isobutylacetophenone, 97%

$174.26 - $669.28

Chemical Identifiers

CAS 38861-78-8
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00027393
InChI Key KEAGRYYGYWZVPC-UHFFFAOYSA-N
Synonym 4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
PubChem CID 93214
IUPAC Name 1-[4-(2-methylpropyl)phenyl]ethanone
SMILES CC(C)CC1=CC=C(C=C1)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1062430 View Documents Alfa Aesar™
A1062430
250g Each for $669.28
 
AAA1062418 View Documents Alfa Aesar™
A1062418
50g Each for $174.26
 
Description

Description

Applications
4′-isobutylacetophenone is an important starting material employed in the production of pharmaceuticals such as ibuprofen. It is also utilized in proteomics research. It is used as an intermediate in organic synthesis.

Solubility
Miscible with chloroform and methanol. Slightly miscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition. Incompatible with reducing agents, strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

38861-78-8
176.259
KEAGRYYGYWZVPC-UHFFFAOYSA-N
93214
CC(C)CC1=CC=C(C=C1)C(=O)C
C12H16O
MFCD00027393
4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
1-[4-(2-methylpropyl)phenyl]ethanone

Specifications

54°C (129°F)
38861-78-8
MFCD00027393
1935275
KEAGRYYGYWZVPC-UHFFFAOYSA-N
1-[4-(2-methylpropyl)phenyl]ethanone
93214
97%
250g
C12H16O
UN1224
4'-isobutylacetophenone, 4-isobutylacetophenone, p-iso-butylacetophenone, 1-4-isobutylphenyl ethanone, p-isobutylacetophenone, ethanone, 1-4-2-methylpropyl phenyl, 1-4-2-methylpropyl phenyl ethan-1-one, acetophenone, 4-isobutyl, 1-4-2-methylpropyl phenyl ethanone, unii-aml715rd20
CC(C)CC1=CC=C(C=C1)C(=O)C
176.259
176.27
4'-Isobutylacetophenone
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA