missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodotriphenylamine, TCI America™

$229.17 - $802.09

Chemical Identifiers

CAS 38257-52-2
Molecular Formula C18H14IN
Molecular Weight (g/mol) 371.221
MDL Number MFCD11046352
InChI Key OWWVTWHBNAWUJO-UHFFFAOYSA-N
Synonym 4-Iodo-N,N-diphenylaniline
PubChem CID 18615936
IUPAC Name 4-iodo-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09511G View Documents TCI America
I09511G
1g Each for $229.17
 
I09515G View Documents TCI America
I09515G
5g Each for $802.09
 
Specifications

Chemical Identifiers

38257-52-2
371.221
OWWVTWHBNAWUJO-UHFFFAOYSA-N
18615936
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)I
C18H14IN
MFCD11046352
4-Iodo-N,N-diphenylaniline
4-iodo-N,N-diphenylaniline

Specifications

Undesignated
1g
C18H14IN
4-Iodo-N,N-diphenylaniline
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)I
371.221
371.22
≥98.0% (GC)
109°C
38257-52-2
MFCD11046352
OWWVTWHBNAWUJO-UHFFFAOYSA-N
4-iodo-N,N-diphenylaniline
18615936
Crystal-Powder at 20°C
4-Iodotriphenylamine
Safety and Handling

Safety and Handling

TSCA : No