Please login to your online account to display your discounted pricing

4-Iodotoluene 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  133941000

 View more versions of this product

Catalog No. AC133941000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Low Melting Mass
Formula Weight 218.04
Quantity 100g
Color White to light yellow
Beilstein 05, 312
EINECS Number 210-841-7
TSCA TSCA
Melting Point 33° to 36°C
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 624-31-7
CAS Min % 97.5
Chemical Name or Material 4-Iodotoluene
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H7I
Linear Formula IC6H4CH3
MDL Number MFCD00001059

Specifications

Chemical Name or Material 4-Iodotoluene
Name Note 98%
CAS 624-31-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7I
Linear Formula IC6H4CH3
Formula Weight 218.04
MDL Number MFCD00001059