missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodopyridine 96%, ACROS Organics™

Chemical Identifiers

CAS 15854-87-2
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C5H4IN
Formula Weight 204.99
MDL Number MFCD02093937
Chemical Name or Material 4-Iodopyridine
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC391290010 SDS Thermo Fisher Scientific
AC391290010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC391290050 SDS Thermo Fisher Scientific
AC391290050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

15854-87-2
100.0
C5H4IN
MFCD02093937
≥95%
95.0
96%
204.99
4-Iodopyridine

Specifications

15854-87-2
100.0
Glass bottle
Crystals or Powder
C5H4IN
MFCD02093937
1g
≥95%
95.0
Authentic
Product darkens on storage
96%
204.99
Pale brown to dark green-brown
4-Iodopyridine

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Freezer -20°C