Learn More
4-Iodopyridine, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 391290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Iodopyridine | |
95.0 | |
Authentic | |
Glass bottle | |
MFCD02093937 | |
pyridine, 4-iodo, 4-iodo pyridine, 4-iodo-pyridine, p-iodopyridine, 4-iodanylpyridine, acmc-1btjc, 4-iodopyridine, ksc178q2d | |
RTLUPHDWSUGAOS-UHFFFAOYSA-N | |
4-iodopyridine | |
609492 | |
96% |
15854-87-2 | |
100.0 | |
95% min. (GC) | |
C5H4IN | |
5g | |
Solubility in water: slightly soluble | |
IC1=CC=NC=C1 | |
205.00 | |
204.99 |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
Recommended Storage : Product darkens on storage
RUO â Research Use Only