Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodopyridine, 96%

$124.68 - $481.80

Chemical Identifiers

Chemical Name or Material 4-Iodopyridine
CAS 15854-87-2
Molecular Formula C5H4IN
Formula Weight 205
MDL Number MFCD02093937
Beilstein 1594
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2545903

  SDS
alfa aesar™
B2545903
1g Each for $124.68

AAB2545906

  SDS
alfa aesar™
B2545906
5g Each for $481.80
Description & Specifications

Specifications

Formula Weight 205
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Iodopyridine
Assay 96%
Molecular Formula C5H4IN
MDL Number MFCD02093937
CAS 15854-87-2
Beilstein 1594
Melting Point 96° to 99°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C