Learn More
4-Iodophenylhydrazine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 399050010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Iodophenylhydrazine | |
94.0 | |
95% | |
C6H7IN2 | |
MFCD00052688 | |
4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine | |
IQMLCMKMSBMMGR-UHFFFAOYSA-N | |
(4-iodophenyl)hydrazine | |
704591 | |
95% |
13116-27-3 | |
100.0 | |
Glass bottle | |
IC6H4NHNH2 | |
1g | |
Solubility in water: insoluble | |
C1=CC(=CC=C1NN)I | |
234.04 | |
234.04 |
Chemical Identifiers
13116-27-3 | |
234.04 | |
IQMLCMKMSBMMGR-UHFFFAOYSA-N | |
704591 | |
C1=CC(=CC=C1NN)I |
C6H7IN2 | |
MFCD00052688 | |
4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine | |
(4-iodophenyl)hydrazine |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.<br/
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fre
GHS Signal Word: Warning
RUO â Research Use Only