Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Iodophenylethynyl)trimethylsilane, 97%

$75.42 - $75.42

Chemical Identifiers

Chemical Name or Material (4-Iodophenylethynyl)trimethylsilane
CAS 134856-58-9
Formula Weight 300.21
MDL Number MFCD04974026
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5525603

  SDS
alfa aesar™
H5525603
1g Each for $75.42
Description & Specifications

Specifications

Formula Weight 300.21
Melting Point 66° to 70°C
Chemical Name or Material (4-Iodophenylethynyl)trimethylsilane
Physical Form Powder
Assay 97%
Sensitivity Light Sensitive
MDL Number MFCD04974026
CAS 134856-58-9
Color White to yellow
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Light sensitive.

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.