missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodophenylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 359480050

 View more versions of this product

Catalog No. AC359480050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Iodophenylboronic acid
96.0
322.0°C to 326.0°C
Authentic
Glass bottle
MFCD01319014
4-iodophenyl boronic acid, 4-iodobenzeneboronic acid, p-iodophenylboronic acid, 4-iodophenylboronicacid, p-iodobenzeneboronic acid, p-iodophenyl boronic acid, boronic acid, 4-iodophenyl, 4-boronoiodobenzene, pubchem6160, pubchem11574
PELJYVULHLKXFF-UHFFFAOYSA-N
(4-iodophenyl)boronic acid
151254
97%
5122-99-6
100.0
Beige
96% min. (HPLC)
C6H6BIO2
5 g
Solubility in water: 25g/L
OB(O)C1=CC=C(I)C=C1
247.83
247.82
Powder

Chemical Identifiers

5122-99-6
247.83
PELJYVULHLKXFF-UHFFFAOYSA-N
151254
OB(O)C1=CC=C(I)C=C1
C6H6BIO2
MFCD01319014
4-iodophenyl boronic acid, 4-iodobenzeneboronic acid, p-iodophenylboronic acid, 4-iodophenylboronicacid, p-iodobenzeneboronic acid, p-iodophenyl boronic acid, boronic acid, 4-iodophenyl, 4-boronoiodobenzene, pubchem6160, pubchem11574
(4-iodophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only