Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodophenylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Iodophenylacetonitrile
CAS 51628-12-7
Molecular Formula C8H6IN
Formula Weight 243.05
MDL Number MFCD00060306
UN Number UN3439
Beilstein 1934479
Synonym 4-Iodobenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1511903

  SDS
alfa aesar™
L1511903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1511906

  SDS
alfa aesar™
L1511906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 243.05
Assay 97%
Synonym 4-Iodobenzyl cyanide
MDL Number MFCD00060306
Chemical Name or Material 4-Iodophenylacetonitrile
Molecular Formula C8H6IN
Health Hazard 1 Danger
CAS 51628-12-7
Melting Point 52° to 54°C
Beilstein 1934479
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
UN Number UN3439
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Flash Point >110°C (230°F)

Applications
4-Iodophenylacetonitrile is used as an intermediate in organic syntheses and in pharmaceuticals.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.