missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodophenylacetic acid, 97%

Manufacturer:  Alfa Aesar™ L1334503

 View more versions of this product

Catalog No. AAL1334503



Description

Description

Applications
4-Iodophenylacetic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in suzuki coupling reaction transport.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

4-Iodophenylacetic acid
1g
C8H7IO2
1940558
FJSHTWVDFAUNCO-UHFFFAOYSA-N
2-(4-iodophenyl)acetic acid
137214
97%
136°C to 140°C
1798-06-7
MFCD00082985
4-iodophenylacetic acid, 2-4-iodophenyl acetic acid, 4-iodophenyl acetic acid, 4-iodophenylaceticacid, p-iodophenylacetic acid, benzeneacetic acid, 4-iodo, 4-iodo-phenyl-acetic acid, pubchem18835, 4-iodoohenylacetic acid, 4-iodobenzeneacetic acid
C1=CC(=CC=C1CC(=O)O)I
262.046
262.05

Chemical Identifiers

1798-06-7
262.046
FJSHTWVDFAUNCO-UHFFFAOYSA-N
137214
C1=CC(=CC=C1CC(=O)O)I
C8H7IO2
MFCD00082985
4-iodophenylacetic acid, 2-4-iodophenyl acetic acid, 4-iodophenyl acetic acid, 4-iodophenylaceticacid, p-iodophenylacetic acid, benzeneacetic acid, 4-iodo, 4-iodo-phenyl-acetic acid, pubchem18835, 4-iodoohenylacetic acid, 4-iodobenzeneacetic acid
2-(4-iodophenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents