Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Iodophenoxyacetic acid, 98+%

$65.73 - $65.73

Chemical Identifiers

Chemical Name or Material 4-Iodophenoxyacetic acid
CAS 1878-94-0
Molecular Formula C8H7IO3
Formula Weight 278.05
MDL Number MFCD00014359
Beilstein 2259999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0119606

  SDS
alfa aesar™
L0119606
5g Each for $65.73
Description & Specifications

Specifications

Formula Weight 278.05
Chemical Name or Material 4-Iodophenoxyacetic acid
Assay ≥98%
Molecular Formula C8H7IO3
MDL Number MFCD00014359
CAS 1878-94-0
Beilstein 2259999
Melting Point 156° to 158°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.