missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodophenol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1345522

 View more versions of this product

Catalog No. AAA1345522



Description

Description

4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as pharmaceutical intermediates, and polarizing films for Liquid Crystal Display (LCD) chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodophenol is a chemical used in chemiluminescence imaging assays and experiments, as diagnostic agents in order to detect cancer cells in patients. Also it is used in the synthesis of agonists towards the estrogen β receptor. Also used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display (LCD) chemicals.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry place in tightly closed container. Store away from acid chlorides and acid anhydrides.
Specifications

Specifications

4-Iodophenol
91°C to 95°C
C6H5IO
100 g
Light sensitive
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
VSMDINRNYYEDRN-UHFFFAOYSA-N
4-iodophenol
10894
220.01
540-38-5
138°C (5 mmHg)
MFCD00002327
1904544
14,5036
Soluble in alcohol and ether. Slightly soluble in water.
OC1=CC=C(I)C=C1
220.01
CHEBI:43521
≥98%

Chemical Identifiers

540-38-5
220.01
VSMDINRNYYEDRN-UHFFFAOYSA-N
10894
4-iodophenol
C6H5IO
MFCD00002327
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
CHEBI:43521
OC1=CC=C(I)C=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 208-745-5

RTECSNumber : SL5600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only