missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodophenetole 98.0+%, TCI America™
$55.92 - $158.44
Chemical Identifiers
| CAS | 699-08-1 |
|---|---|
| Molecular Formula | C8H9IO |
| Molecular Weight (g/mol) | 248.063 |
| MDL Number | MFCD00061124 |
| InChI Key | VSIIHWOJPSSIDI-UHFFFAOYSA-N |
| Synonym | 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether |
| PubChem CID | 220700 |
| IUPAC Name | 1-ethoxy-4-iodobenzene |
| SMILES | CCOC1=CC=C(C=C1)I |
Chemical Identifiers
| 699-08-1 | |
| 248.063 | |
| VSIIHWOJPSSIDI-UHFFFAOYSA-N | |
| 220700 | |
| CCOC1=CC=C(C=C1)I |
| C8H9IO | |
| MFCD00061124 | |
| 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether | |
| 1-ethoxy-4-iodobenzene |
Specifications
| 699-08-1 | |
| MFCD00061124 | |
| 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether | |
| CCOC1=CC=C(C=C1)I | |
| 248.063 | |
| 248.06 | |
| Crystalline Lumps |
| C8H9IO | |
| 5 g | |
| VSIIHWOJPSSIDI-UHFFFAOYSA-N | |
| 1-ethoxy-4-iodobenzene | |
| 220700 | |
| ≥98.0% (GC) | |
| 4-Iodophenetole |
Safety and Handling
TSCA : No