Learn More
4-(Iodomethyl)tetrahydro-2h-pyran, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC29940DA
Specifications
4-(Iodomethyl)tetrahydro-2H-pyran | |
97% | |
C6H11IO | |
4-iodomethyl tetrahydro-2h-pyran, 4-iodomethyl oxane, 4-iodomethyl-tetrahydro-pyran, 4-iodomethyl tetrahydropyran, 2h-pyran, tetrahydro-4-iodomethyl, 4-iodomethyl-tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-iodomethyl, acmc-20a1ce, 4-iodomethyltetrahydropyran, 4-iodomethyl-tetrahydropyran | |
C1COCCC1CI | |
226.057 | |
226.05 |
101691-94-5 | |
Amber Glass Bottle | |
1 g | |
MQLFSPBSNWUXSO-UHFFFAOYSA-N | |
4-(iodomethyl)oxane | |
2795507 | |
97% |
Chemical Identifiers
101691-94-5 | |
226.057 | |
4-iodomethyl tetrahydro-2h-pyran, 4-iodomethyl oxane, 4-iodomethyl-tetrahydro-pyran, 4-iodomethyl tetrahydropyran, 2h-pyran, tetrahydro-4-iodomethyl, 4-iodomethyl-tetrahydro-2h-pyran, 2h-pyran,tetrahydro-4-iodomethyl, acmc-20a1ce, 4-iodomethyltetrahydropyran, 4-iodomethyl-tetrahydropyran | |
4-(iodomethyl)oxane |
C6H11IO | |
MQLFSPBSNWUXSO-UHFFFAOYSA-N | |
2795507 | |
C1COCCC1CI |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger