Learn More
4-Iodobenzyl bromide, 97%, Thermo Scientific Chemicals
$57.37 - $163.32
Chemical Identifiers
CAS | 16004-15-2 |
---|---|
Molecular Formula | C7H6BrI |
Molecular Weight (g/mol) | 296.93 |
InChI Key | BQTRMYJYYNQQGK-UHFFFAOYSA-N |
Synonym | 4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide |
PubChem CID | 954258 |
IUPAC Name | 1-(bromomethyl)-4-iodobenzene |
SMILES | C1=CC(=CC=C1CBr)I |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC342460010
|
Thermo Scientific Chemicals
342460010 |
1 g | Glass bottle |
Each for $57.37
|
|
||||
AC342460050
|
Thermo Scientific Chemicals
342460050 |
5 g | Glass bottle |
Each for $163.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16004-15-2 | |
296.93 | |
4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide | |
1-(bromomethyl)-4-iodobenzene |
C7H6BrI | |
BQTRMYJYYNQQGK-UHFFFAOYSA-N | |
954258 | |
C1=CC(=CC=C1CBr)I |
Specifications
16004-15-2 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
IC6H4CH2Br | |
4-iodobenzyl bromide, 1-bromomethyl-4-iodobenzene, 4-iodobenzylbromide, p-iodobenzyl bromide, 1-bromomethyl-4-iodo-benzene, alpha-bromo-4-iodotoluene, 4-bromomethyl-1-iodobenzene, benzene, 1-bromomethyl-4-iodo, 4-iodo benzyl bromide | |
C1=CC(=CC=C1CBr)I | |
296.93 | |
296.93 | |
Adhering Crystalline Powder |
96.0 | |
77.0°C to 82.0°C | |
140.0°C (15.0 mmHg) | |
96% min. (GC) | |
C7H6BrI | |
1 g | |
BQTRMYJYYNQQGK-UHFFFAOYSA-N | |
1-(bromomethyl)-4-iodobenzene | |
954258 | |
97% | |
4-Iodobenzyl bromide |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only