Learn More
4-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals
$129.44 - $1,227.19
Chemical Identifiers
CAS | 1711-02-0 |
---|---|
Molecular Formula | C7H4ClIO |
Molecular Weight (g/mol) | 266.462 |
MDL Number | MFCD00001058 |
InChI Key | NJAKCIUOTIPYED-UHFFFAOYSA-N |
Synonym | p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride |
PubChem CID | 74373 |
IUPAC Name | 4-iodobenzoyl chloride |
SMILES | C1=CC(=CC=C1C(=O)Cl)I |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2538806
|
Thermo Scientific Chemicals
B2538806 |
5 g |
N/A
|
|
|||||
AAB2538814
|
Thermo Scientific Chemicals
B2538814 |
25 g |
N/A
|
|
|||||
AAB2538822
|
Thermo Scientific Chemicals
B2538822 |
100 g |
N/A
|
|
|||||
Description
4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Iodobenzoyl chloride is employed in the preparation of pyrroles by reaction with imines and acetylenes mediated by isocyanides using palladium catalysis. It is also used in the preparation of N-(1-benzylpyrrolidin-3-yl)arylbenzamides, potential human dopamine D4 antagonists. Further, it is used to prepare [2]rotaxane monomer and poly[2] rotaxane. In addition to this, it is important in the modification of poly(allylamine) in order to make the polymer X-ray visible.
Solubility
Soluble in toluene and benzene.
Notes
Moisture sensitive. Incompatible with strong bases and water.
Chemical Identifiers
1711-02-0 | |
266.462 | |
NJAKCIUOTIPYED-UHFFFAOYSA-N | |
74373 | |
C1=CC(=CC=C1C(=O)Cl)I |
C7H4ClIO | |
MFCD00001058 | |
p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride | |
4-iodobenzoyl chloride |
Specifications
1711-02-0 | |
120°C to 121°C (1 mmHg) | |
C7H4ClIO | |
5 g | |
774718 | |
p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride | |
NJAKCIUOTIPYED-UHFFFAOYSA-N | |
4-iodobenzoyl chloride | |
74373 | |
98% |
61°C to 65°C | |
>110°C (230°F) | |
MFCD00001058 | |
UN3261 | |
Light and moisture sensitive | |
Soluble in toluene and benzene. | |
C1=CC(=CC=C1C(=O)Cl)I | |
266.462 | |
266.47 | |
4-Iodobenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 216-974-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only