missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Iodobenzoic acid, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 124480100

 View more versions of this product

Catalog No. AC124480100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Iodobenzoic acid
Authentic
White
10g
619-58-9
100.0
C7H5IO2
MFCD00002533
Solubility in water: 0.04g/L (25°C)
C1=CC(=CC=C1C(=O)O)I
248.02
248.02
98%
>110°C
Glass bottle
270°C to 274°C
98%
97.5
97.5% min. (HPLC)
IC6H4COOH
4-iodo benzoic acid, 4-iodo-benzoic acid, 4-jodbenzoesaeure, 4-jodbenzoesaeure german, benzoic acid, 4-iodo, benzoic acid, p-iodo, ipo4lyq1en, p-iodobenzenecarboxylic acid, p-iodobenzoic acid, unii-ipo4lyq1en
GHICCUXQJBDNRN-UHFFFAOYSA-N
4-iodobenzoic acid
12085
Powder

Chemical Identifiers

619-58-9
248.02
GHICCUXQJBDNRN-UHFFFAOYSA-N
12085
C1=CC(=CC=C1C(=O)O)I
C7H5IO2
MFCD00002533
4-iodo benzoic acid, 4-iodo-benzoic acid, 4-jodbenzoesaeure, 4-jodbenzoesaeure german, benzoic acid, 4-iodo, benzoic acid, p-iodo, ipo4lyq1en, p-iodobenzenecarboxylic acid, p-iodobenzoic acid, unii-ipo4lyq1en
4-iodobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-603-2

RTECSNumber : DH2976000

TSCA : TSCA

SDS Documents